missing translation for 'onlineSavingsMsg'
Learn More

Diphenmanil methylsulfate, MedChemExpress

Product Code. 16446998
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446998 10 mg 10mg
16403057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446998 Supplier Medchem Express Supplier No. HY16171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Diphemanil methylsulfate is a quaternary ammonium anticholinergic.

Diphemanil methylsulfate is a quaternary ammonium anticholinergic.

Specifications

Chemical Name or Material Diphenmanil methylsulfate
Physical Form Solid
CAS 62-97-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H27NO4S
Synonym Diphemanil mesylate
Solubility Information H2O : 16.67 mg/mL (42.80 mM; Need ultrasonic)
SMILES [O-]S(=O)(OC)=O.C[N+]1(C)CC/C(CC1)=C(C2=CC=CC=C2)\C3=CC=CC=C3
Molecular Weight (g/mol) 389.51
Formula Weight 389.51
Percent Purity 99.74%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.