missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Diphenhydramine hydrochloride

Product Code. 18772714
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772714 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772714 Supplier Tocris Bioscience™ Supplier No. 3072/50
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

H1 receptor antagonist

H1 receptor antagonist. Antihistamine.
TRUSTED_SUSTAINABILITY

Specifications

CAS 147-24-0
Quantity 50 mg
Molecular Formula C17H22ClNO
Formula Weight 291.82
Purity >99%
Molecular Weight (g/mol) 291.82
Chemical Name or Material Diphenhydramine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.