missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenhydramine Hydrochloride, Mikromol™
Descrizione
Diphenhydramine Hydrochloride
Specifica
Specifica
| Chemical Name or Material | Diphenhydramine hydrochloride |
| CAS | 147-24-0 |
| Analyte or Component Names | Diphenhydramine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H21 N O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Orphenadrine Hydrochloride Imp. D (EP), Orphenadrine Imp. D (EP), Diphenhydramine Hydrochloride, Orphenadrine Hydrochloride Imp. D (EP) as Hydrochloride, Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (9CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8CI), b-Dimethylaminoethyl benzhydryl ether hydrochloride, 2-(Benzhydryloxy)-N,N-dimethylethylamine hydrochloride, 2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, Alledryl, Allergan, Allergina, Allergival, Amidryl, Asdrin, Bagodryl, Bax, Bena, Bena-Fedrin, Benadril hydrochloride, Benadryl, Benocten, Benodine, Benylin, Benzantin, Benzhydramine hydrochloride, Benzhydrol b-dimethylaminoethyl ether hydrochloride, Denydryl, Dibondrin, Difenhydramine hydrochloride, Dimedrol, Dimedrol hydrochloride, Diphantine, Fenylhist, Halbmond, Histacyl, Noctomin, Resmin, Restamin, S 8, Sedopretten, Sekundal D, Syntedril, Vena, Wehydryl, Orphenadrine Hydrochloride Impurity D as Hydrochloride, Orphenadrine Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCOC(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?