missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine hydrochloride, MedChemExpress

Product Code. 16450868
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 mg
5 g
Unit Size:
250mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450868 5 g 5g
16429875 250 mg 250mg
16422096 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16450868 Supplier Medchem Express Supplier No. HYB0303A
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with anti-cholinergic effect. Diphenhydramine hydrochloride can across the ovine blood-brain barrier (BBB).

Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with anti-cholinergic effect. Diphenhydramine hydrochloride can across the ovine blood-brain barrier (BBB).

Specifications

Chemical Name or Material Diphenhydramine hydrochloride
Physical Form Solid
CAS 147-24-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C17H22ClNO
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (342.68 mM; Need ultrasonic) ∣H2O : 100 mg/mL (342.68 mM; Need ultrasonic)
SMILES CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2.Cl
Molecular Weight (g/mol) 291.82
Formula Weight 291.82
Percent Purity 99.04%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.