missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number 10193581
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
147-24-0
C17H22ClNO
291.82
MFCD00012479
PCHPORCSPXIHLZ-UHFFFAOYSA-N
diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
8980
CHEBI:4637
2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
147-24-0
C17H22ClNO
291.82
MFCD00012479
PCHPORCSPXIHLZ-UHFFFAOYSA-N
diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
8980
CHEBI:4637
2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 168°C to 172°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 3339
Solubility Information Solubility in water: 1000g/L. Other solubilities: 500g/L ethanol
Formula Weight 291.82
Percent Purity 99%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material Diphenhydramine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diphenhydramine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'