missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine Citrate, TRC

Product Code. 30387235
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387235 250 mg -
30371304 1 g 1g
30386165 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387235 Supplier Toronto Research Chemicals Supplier No. TRCD486905250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88637-37-0
Molecular Formula C17 H21 N O . C6 H8 O7
Molecular Weight (g/mol) 447.48
Synonym Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Diphenhydramine citrate
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CN(C)CCOC(c1ccccc1)c2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H21NO.C6H8O7/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-12,17H,13-14H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 447.1893
Chemical Name or Material Diphenhydramine Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.