missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphemanil Metilsulfate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Diphemanil Metilsulfate |
| CAS | 62-97-5 |
| Analyte or Component Names | Diphemanil Metilsulfate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N . C H3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Diphemanil Metilsulfate, 4-(Diphenylmethylene)-1,1-dimethylpiperidinium methyl sulfate, Demotil, Diphemanil, Diphemanil methosulfate, Diphemanil methyl sulfate, Diphemanil metilsulfate, Diphenatil, Diphenmanil methylsulfate, Diphenmethanil, Diphenmethanil methylsulfate, N,N-Dimethyl-4-piperidylidene-1,1-diphenylmethane methylsulfate, NSC 41725, Nivelon, Nivelona, Prantal, Prantil, Prentol, Talpran, Vagophemanil, Vagophemanil methyl sulfate, Variton |
| InChI Formula | InChI=1S/C20H24N.CH4O4S/c1-21(2)15-13-19(14-16-21)20(17-9-5-3-6-10-17)18-11-7-4-8-12-18;1-5-6(2,3)4/h3-12H,13-16H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1 |
| Solution Type | Neat |
| SMILES | COS(=O)(=O)[O-].C[N+]1(C)CCC(=C(c2ccccc2)c3ccccc3)CC1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?