missing translation for 'onlineSavingsMsg'
Learn More

Diphemanil Methosulfate, TRC

Product Code. 30382753
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382753 100 mg
30384631 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382753 Supplier Toronto Research Chemicals Supplier No. TRCD486965100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-97-5
Molecular Formula C20 H24 N . C H3 O4 S
Molecular Weight (g/mol) 389.51
Synonym Diphemanil Metilsulfate, 4-(Diphenylmethylene)-1,1-dimethylpiperidinium methyl sulfate, Demotil, Diphemanil, Diphemanil methosulfate, Diphemanil methyl sulfate, Diphemanil metilsulfate, Diphenatil, Diphenmanil methylsulfate, Diphenmethanil, Diphenmethanil methylsulfate, N,N-Dimethyl-4-piperidylidene-1,1-diphenylmethane methylsulfate, NSC 41725, Nivelon, Nivelona, Prantal, Prantil, Prentol, Talpran, Vagophemanil, Vagophemanil methyl sulfate, Variton
IUPAC Name 4-benzhydrylidene-1,1-dimethylpiperidin-1-ium;methyl sulfate
SMILES COS(=O)(=O)[O-].C[N+]1(C)CCC(=C(c2ccccc2)c3ccccc3)CC1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H24N.CH4O4S/c1-21(2)15-13-19(14-16-21)20(17-9-5-3-6-10-17)18-11-7-4-8-12-18;1-5-6(2,3)4/h3-12H,13-16H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1
Formula Weight 389.1661
Chemical Name or Material Diphemanil Metilsulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.