missing translation for 'onlineSavingsMsg'
Learn More

Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals

Catalog Number 10021050
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
126-58-9
C10H22O7
254.28
MFCD00004691
TXBCBTDQIULDIA-UHFFFAOYSA-N
dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol
31352
2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)COCC(CO)(CO)CO)O
This item is not returnable. View return policy
126-58-9
C10H22O7
254.28
MFCD00004691
TXBCBTDQIULDIA-UHFFFAOYSA-N
dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol
31352
2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)COCC(CO)(CO)CO)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 215°C to 225°C
Color White
Boiling Point 356°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Plastic bottle
Linear Formula (HOCH2)3CCH2OCH2C(CH2OH)3
Quantity 500 g
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: 0.99g/100 mL water (50°C),2.1g/L methanol (25°C),0.6g/L ethanol (25°C)
Formula Weight 254.28
Percent Purity 85+%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Dipentaerythritol, 90%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'