missing translation for 'onlineSavingsMsg'
Learn More

Dipentaerythritol, 80+%, technical

Product Code. 10070810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070810 25 g Plastic bottle 25g
10021050 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10070810 Supplier Thermo Scientific Acros Supplier No. 117120250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126-58-9 | C10H22O7 | 254.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-58-9
Molecular Formula C10H22O7
Molecular Weight (g/mol) 254.28
MDL Number MFCD00004691
InChI Key TXBCBTDQIULDIA-UHFFFAOYSA-N
Synonym dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol
PubChem CID 31352
IUPAC Name 2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)COCC(CO)(CO)CO)O

Specifications

Melting Point 215°C to 225°C
Color White
Boiling Point 356°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Plastic bottle
Linear Formula (HOCH2)3CCH2OCH2C(CH2OH)3
Quantity 25 g
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: 0.99g/100 mL water (50°C),2.1g/L methanol (25°C),0.6g/L ethanol (25°C)
Formula Weight 254.28
Percent Purity 85+%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Dipentaerythritol, 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.