missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563241
This item is not returnable.
View return policy
Description
Immunogen sequence: IGEICVSSRT GGMMYFGLAG VTKNTFEVIP VNSAGSPVGD VPFIRSGLL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes a member of the disco-interacting protein homolog 2 protein family. The encoded protein contains a binding site for the transcriptional regulator DNA methyltransferase 1 associated protein 1 as well as AMP-binding sites. The presence of these sites suggests that the encoded protein may participate in DNA methylation. This gene is located near a folate-sensitive fragile site, and CGG-repeat expansion in the promoter of this gene which affects transcription has been detected in individuals containing this fragile site on chromosome 12.
Specifications
| DIP2B | |
| Polyclonal | |
| Unconjugated | |
| DIP2B | |
| 4932422C22; AI317237; AI854602; DIP2 disco-interacting protein 2 homolog B; DIP2 disco-interacting protein 2 homolog B (Drosophila); DIP2 homolog B; Dip2b; disco interacting protein 2 homolog B; disco-interacting protein 2 homolog B; KIAA1463; mKIAA1463 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57609 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P265 | |
| DIP2B | |
| Recombinant protein corresponding to Human DIP2B. Recombinant protein control fragment (Product #RP-103243). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction