missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIP13B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558884
This item is not returnable.
View return policy
Description
Immunogen sequence: PMIGFAHGQI NFFKKGAEMF SKRMDSFLSS VADMVQSIQV ELEAEAEKMR VSQQELLSVD ESVYTPDSDV AAPQINRNLI QK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.
Specifications
| DIP13B | |
| Polyclonal | |
| Unconjugated | |
| APPL2 | |
| adapter protein containing PH domain, PTB domain and leucine zipper motif 2; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2; Appl2; DCC-interacting protein 13-beta; DIP13 beta; DIP13B; dip13-beta; Dip3 beta; Dip3b; RGD1563028 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55198 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEU8 | |
| APPL2 | |
| Recombinant protein corresponding to Human DIP13B. Recombinant protein control fragment (Product #RP-97755). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction