missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DIP13B Antibody, Novus Biologicals™

Product Code. 18347109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347109 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347109 Supplier Novus Biologicals™ Supplier No. H00055198B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

DIP13B Polyclonal antibody specifically detects DIP13B in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIP13B
Applications Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. AAH33731.1
Gene Alias Adapter protein containing PH domain, PTB domain and leucine zipper motif 2, adaptor protein containing PH domain, PTB domain and leucine zipper motif 2, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 2, DCC-interacting protein 13-beta, DIP13BDIP13 beta, Dip13-beta, FLJ10659dip13-beta
Host Species Mouse
Immunogen APPL2 (AAH33731.1, 1 a.a. - 664 a.a.) full-length human protein. MPAVDKLLLEEALQDSPQTRSLLSVFEEDAGTLTDYTNQLLQAMQRVYGAQNEMCLATQQLSKQLLAYEKQNFALGKGDEEVISTLHYFSKVVDELNLLHTELAKQLADTMVLPIIQFREKDLTEVSTLKDLFGLASNEHDLSMAKYSRLPKKKENEKVKTEVGKEVAAARRKQHLSSLQYYCALNALQYRKQMAMMEPMIGFAHGQINFFKKGAEMFSKRMDSFLSSVADMVQSIQVELEAEAEKMRVSQQELLSVDESVYTPDSDVAAPQINRNLIQKAGYLNLRNKTGLVTTTWERLYFFTQGGNLMCQPRGAVAGGLIQDLDNCSVMAVDCEDRRYCFQITTPNGKSGIILQAESRKENEEWICAINNISRQIYLTDNPEAVAIKLNQTALQAVTPITSFGKKQESSCPSQNLKNSEMENENDKIVPKVTASLPEAEELIAPGTPIQFDIVLPATEFLDQNRGSRRTNPFGETEDESFPEAEDSLLQQMFIVRFLGSMAVKTDSTTEVIYEAMRQVLAARAIHNIFRMTESHLMVTSQSLRLIDPQTQVSRANFELTSVTQFAAHQENKRLVGFVIRVPESTGEESLSTYIFESNSEGEKICYAINLGKEIIEVQKDPEALAQLMLSIPLTNDGKYVLLNDQPDDDDGNPNEHRGAESEA
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55198
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.