Learn More
Fistreem International™ Dip Tube (Boiler), Loughborough Water Still
Click to view available options
For Use With (Equipment):
WS/4 Water Still
WS/8 Water Still
Unit Size:
Each
Description
- For Loughborough water still.
- Directs incoming cold replacement water to the bottom of the boiler.
- Includes silicone rubber tip.
Specifications
Specifications
| Includes | Silicone Rubber Tip |
| Material | Silicone Rubber |
| For Use With (Equipment) | WS/8 Water Still |
| Type | Dip Tube |