Learn More
Fistreem International™ Dip Tube (Boiler), Loughborough Water Still
Product Code. 13156000
Click to view available options
For Use With (Equipment):
WS/4 Water Still
WS/8 Water Still
Unit Size:
Each
Product Code. 13156000
Brand: Fistreem International™ ALB/2/4
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifically designed for a Loughborough water still. Fistreem International™ Dip Tube (Boiler), Loughborough Water Still directs incoming cold replacement water to the bottom of the boiler.
  • For Loughborough water still.
  • Directs incoming cold replacement water to the bottom of the boiler.
  • Includes silicone rubber tip.
Specifications
Includes Silicone Rubber Tip
For Use With (Equipment) WS/4 Water Still
Type Dip Tube