missing translation for 'onlineSavingsMsg'
Learn More

Dioxidine, TRC

Product Code. 17947164
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17947164 50 mg 50mg
17909264 5 mg 5mg
17918374 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17947164 Supplier Toronto Research Chemicals Supplier No. TRCD48716050MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17311-31-8
Molecular Formula C10 H10 N2 O4
Molecular Weight (g/mol) 222.197
Synonym 2,3-Bis(hydroxymethyl)quinoxaline-1,4-di-N-oxide, 2,3-Di(hydroxymethyl)quinoxaline 1,4-Di-N-oxide, 2,3-Di(hydroxymethyl)quinoxaline Di-N-oxide, Dioxydine, Farmoxidin, Mastoxidine, Metrosept;
IUPAC Name [3-(hydroxymethyl)-1,4-dioxo-quinoxalin-2-yl]methanol
SMILES OCc1c(CO)n(=O)c2ccccc2n1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H10N2O4/c13-5-9-10(6-14)12(16)8-4-2-1-3-7(8)11(9)15/h1-4,13-14H,5-6H2
Formula Weight 222.0641
Percent Purity >95%
Chemical Name or Material Dioxidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.