missing translation for 'onlineSavingsMsg'
Learn More

Dioxethedrine Hydrobromide, TRC

Product Code. 30384760
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384760 50 mg 50mg
30383518 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384760 Supplier Toronto Research Chemicals Supplier No. TRCD48623050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dioxethedrine Hydrobromide
Analyte or Component Names Dioxethedrine Hydrobromide
Shipping Condition Room Temperature
Molecular Formula C11 H17 N O3 . HBr
Synonym 4-(2-(Ethylamino)-1-hydroxypropyl)benzene-1,2-diol Hydrobromide
InChI Formula InChI=1S/C11H17NO3.BrH/c1-3-12-7(2)11(15)8-4-5-9(13)10(14)6-8;/h4-7,11-15H,3H2,1-2H3;1H
Solution Type Neat
SMILES Br.CCNC(C)C(O)c1ccc(O)c(O)c1
IUPAC Name 4-[2-(ethylamino)-1-hydroxypropyl]benzene-1,2-diol;hydrobromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 292.17
Quantity 50 mg
Formula Weight 291.047
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.