missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dioxane R1 solution EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Packaging:
Ampoule
Specifications
Specifications
CAS | 6988-21-2 |
Packaging | Ampoule |
Quantity | 100 mL |
Type | Pesticide and Metabolite Standards |
Molecular Formula | C11H13NO4 |
InChI Key | SDKQRNRRDYRQKY-UHFFFAOYSA-N |
SMILES | CNC(=O)OC1=CC=CC=C1C2OCCO2 |
IUPAC Name | [2-(1,3-dioxolan-2-yl)phenyl] N-methylcarbamate |
Molecular Weight (g/mol) | 223.228 |
PubChem CID | 23421 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'