missing translation for 'onlineSavingsMsg'
Learn More

Diosmin Octaacetate, TRC

Product Code. 30384231
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384231 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384231 Supplier Toronto Research Chemicals Supplier No. TRCD4852051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diosmin Octaacetate
CAS 6195-54-6
Molecular Formula C44H48O23
Quantity 1 g
Synonym 5-(Acetyloxy)-2-[3-(acetyloxy)-4-methoxyphenyl]-7-[[2,3,4-tri-O-acetyl-6-O-(2,3,4-tri-O-acetyl-6-deoxy-alpha-L-mannopyranosyl)-beta-D-glucopyranosyl]oxy]-4H-1-benzopyran-4-one
InChI Formula InChI=1S/C44H48O23/c1-18-37(59-21(4)47)39(61-23(6)49)41(63-25(8)51)43(56-18)55-17-35-38(60-22(5)48)40(62-24(7)50)42(64-26(9)52)44(67-35)65-28-14-33(58-20(3)46)36-29(53)16-31(66-34(36)15-28)27-11-12-30(54-10)32(13-27)57-19(2)45/h11-16,18,35,37-44H,17H2,1-10H3/t18-,35?,37+,38+,39+,40?,41+,42+,43+,44+/m0/s1
SMILES COc1ccc(cc1OC(=O)C)C2=CC(=O)c3c(OC(=O)C)cc(O[C@@H]4OC(CO[C@@H]5O[C@@H](C)[C@@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]5OC(=O)C)[C@@H](OC(=O)C)C(OC(=O)C)[C@H]4OC(=O)C)cc3O2
IUPAC Name [5-[5-acetyloxy-4-oxo-7-[(2S,3R,5R)-3,4,5-triacetyloxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-triacetyloxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-2-yl]-2-methoxyphenyl] acetate
Molecular Weight (g/mol) 944.84
Formula Weight 944.2586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.