missing translation for 'onlineSavingsMsg'
Learn More

Diosmetine Triacetate, TRC

Product Code. 30371190
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371190 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371190 Supplier Toronto Research Chemicals Supplier No. TRCD4850302MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diosmetine Triacetate
CAS 3162-05-8
Molecular Formula C22H18O9
Quantity 2 mg
Synonym 5,7-Bis(acetyloxy)-2-[3-(acetyloxy)-4-methoxyphenyl]-4H-1-benzopyran-4-one, 3',5,7-Trihydroxy-4'-methoxy-flavone Triacetate, 3',5,7-Triacetoxy-4'-methoxyflavone, Diosmetin Triacetate
InChI Formula InChI=1S/C22H18O9/c1-11(23)28-15-8-20(30-13(3)25)22-16(26)10-18(31-21(22)9-15)14-5-6-17(27-4)19(7-14)29-12(2)24/h5-10H,1-4H3
SMILES COc1ccc(cc1OC(=O)C)C2=CC(=O)c3c(OC(=O)C)cc(OC(=O)C)cc3O2
IUPAC Name [5-acetyloxy-2-(3-acetyloxy-4-methoxyphenyl)-4-oxochromen-7-yl] acetate
Molecular Weight (g/mol) 426.37
Formula Weight 426.0951
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.