missing translation for 'onlineSavingsMsg'
Learn More

Diosmetin 3’,7-Diglucuronide (>85%), TRC

Product Code. 30383101
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383101 1 mg 1mg
30371189 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383101 Supplier Toronto Research Chemicals Supplier No. TRCD4850401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152503-51-0
Molecular Formula C28 H28 O18
Molecular Weight (g/mol) 652.51
Synonym β-D-Glucopyranosiduronic acid, 5-[7-(β-D-glucopyranuronosyloxy)-5-hydroxy-4-oxo-4H-1-benzopyran-2-yl]-2-methoxyphenyl
IUPAC Name (2S,3S,4S,5R,6R)-6-[2-[3-[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy-4-methoxyphenyl]-5-hydroxy-4-oxochromen-7-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES COc1ccc(cc1O[C@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)C3=CC(=O)c4c(O)cc(O[C@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)cc4O3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C28H28O18/c1-41-12-3-2-8(4-14(12)44-28-22(36)18(32)20(34)24(46-28)26(39)40)13-7-11(30)16-10(29)5-9(6-15(16)43-13)42-27-21(35)17(31)19(33)23(45-27)25(37)38/h2-7,17-24,27-29,31-36H,1H3,(H,37,38)(H,39,40)/t17-,18-,19-,20-,21+,22+,23-,24-,27-,28-/m0/s1
Formula Weight 652.1276
Chemical Name or Material Diosmetin 3',7-Diglucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.