missing translation for 'onlineSavingsMsg'
Learn More

Diosgenin

Product Code. 15473649
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15473649

Brand: Thermo Scientific Alfa Aesar J60976.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 512-04-9 | C27H42O3 | 414.63 g/mol

Diosgenin is the precursor for the semisynthesis of progesterone which in turn was used in early combined oral contraceptive pills. The unmodified steroid has estrogenic activity and can reduce the level of serum cholesterol. Diosgenin has been used in traditional medicine as an antihypercholesterolemia, antihypertriacylglycerolemia, antidiabetes and antihyperglycemia agent. Diosgenin is a steroid sapogenin used in the semi-synthesis of pregnenolone, cortisone, and progesterone. Diosgenin has estrogenic activity and can reduce the level of serum cholesterol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diosgenin is the precursor for the semisynthesis of progesterone which in turn was used in early combined oral contraceptive pills. The unmodified steroid has estrogenic activity and can reduce the level of serum cholesterol. Diosgenin has been used in traditional medicine as an antihypercholesterolemia, antihypertriacylglycerolemia, antidiabetes and antihyperglycemia agent. Diosgenin is a steroid sapogenin used in the semi-synthesis of pregnenolone, cortisone, and progesterone. Diosgenin has estrogenic activity and can reduce the level of serum cholesterol.

Solubility
Soluble in chloroform (50 mg/ml), ethanol (83 mg/ml at 25°C), water (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-04-9
Molecular Formula C27H42O3
Molecular Weight (g/mol) 414.63
MDL Number MFCD00016887,MFCD01002142
InChI Key WQLVFSAGQJTQCK-VKROHFNGSA-N
Synonym 3-beta-Hydroxy-5-spirostene; Nitogenin
PubChem CID 46783391
IUPAC Name (1'S,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'R,16'S)-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-18'-en-16'-ol
SMILES C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C)O[C@]11CC[C@@H](C)CO1

Specifications

Melting Point 200°C to 206°C
Color White
Quantity 25 g
Beilstein 94582
Merck Index 14,3295
Solubility Information Soluble in chloroform (50mg/ml),ethanol (83mg/ml at 25°C),water (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),and methanol.
Formula Weight 414.63
Physical Form Powder
Chemical Name or Material Diosgenin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.