missing translation for 'onlineSavingsMsg'
Learn More

Diosgenin, TRC

Product Code. 30138500
Change view
Click to view available options
Quantity:
250 mg
5 g
10 g
Unit Size:
10g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138500 5 g 5g
30006883 10 g 10g
30012778 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138500 Supplier Toronto Research Chemicals Supplier No. TRCD4847005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-04-9
Molecular Formula C27 H42 O3
Molecular Weight (g/mol) 414.62
Synonym Spirost-5-en-3-ol, (3β,25R)-, 25D-Spirost-5-en-3β-ol (6CI,7CI), Spirost-5-en-3β-ol, (25R)- (8CI), Spiro[8H-naphth[2',1':4,5]indeno[2,1-b]furan-8,2'-[2H]pyran], spirost-5-en-3-ol deriv., (25R)-Spirost-5-en-3β-ol, (3β,25R)-Spirost-5-en-3-ol, 25α-Spirost-5-en-3β-ol, Diosgenin, NSC 33396, Nitogenin
SMILES C[C@@H]1CC[C@@]2(OC1)O[C@H]3C[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]4(C)[C@H]3[C@@H]2C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C27H42O3/c1-16-7-12-27(29-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(28)8-10-25(18,3)21(20)9-11-26(22,24)4/h5,16-17,19-24,28H,6-15H2,1-4H3/t16-,17+,19+,20-,21+,22+,23+,24+,25+,26+,27-/m1/s1
Formula Weight 414.3134
Percent Purity >95%
Chemical Name or Material Diosgenin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.