Learn More
Diosgenin
Description
Diosgenin is the precursor for the semisynthesis of progesterone which in turn was used in early combined oral contraceptive pills. The unmodified steroid has estrogenic activity and can reduce the level of serum cholesterol. Diosgenin has been used in traditional medicine as an antihypercholesterolemia, antihypertriacylglycerolemia, antidiabetes and antihyperglycemia agent. Diosgenin is a steroid sapogenin used in the semi-synthesis of pregnenolone, cortisone, and progesterone. Diosgenin has estrogenic activity and can reduce the level of serum cholesterol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diosgenin is the precursor for the semisynthesis of progesterone which in turn was used in early combined oral contraceptive pills. The unmodified steroid has estrogenic activity and can reduce the level of serum cholesterol. Diosgenin has been used in traditional medicine as an antihypercholesterolemia, antihypertriacylglycerolemia, antidiabetes and antihyperglycemia agent. Diosgenin is a steroid sapogenin used in the semi-synthesis of pregnenolone, cortisone, and progesterone. Diosgenin has estrogenic activity and can reduce the level of serum cholesterol.
Solubility
Soluble in chloroform (50 mg/ml), ethanol (83 mg/ml at 25°C), water (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 200°C to 206°C |
| Color | White |
| Quantity | 5 g |
| Beilstein | 94582 |
| Merck Index | 14,3295 |
| Solubility Information | Soluble in chloroform (50mg/ml),ethanol (83mg/ml at 25°C),water (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),and methanol. |
| Formula Weight | 414.63 |
| Physical Form | Powder |
| Chemical Name or Material | Diosgenin |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.