missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific™ Dionex Protein Mixture Digest is lyophilized.
Specifications
Specifications
| Description | Protein mixture digest |
| For Use With (Equipment) | Acclaim PepMap C18 Column |
| Type | Protein Mixture Digest |
| Standard Components | Tryptic peptides, Trypsin, Ammonium bicarbonate, Trifluoroacetic acid |
| Standard Concentration | 100 pmol |
| Storage | -20°C |
| Product Line | Dionex |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?