Learn More
Thermo Scientific™ Dionex™ PEEK Sleeves
Product Code. 11372133 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Connections with Fused Silica Tubing
Connections with Microtight Union
Diameter (Metric) Outer:
30 μm
280 μm
360 μm
Unit Size:
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Outer unitSize
11372133 Connections with Fused Silica Tubing 280 μm Pack of 5
11382133 Connections with Fused Silica Tubing 30 μm Pack of 5
11382163 Connections with Microtight Union 360 μm Pack of 10
11352173 Connections with Microtight Union 280 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11372133 Supplier Thermo Scientific™ Supplier No. 160492
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
TRUSTED_SUSTAINABILITY
Specifications
Description For fused silica tubing; 280 um OD
For Use With (Equipment) Connections with Fused Silica Tubing
Type Tubing Sleeve
Diameter (Metric) Outer 280 μm
Product Line nanoLC