missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OmniPac™ PCX-500 HPLC Columns

Product Code. 11331723 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
50 mm
250 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11331723 250 mm Each
11351723 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331723 Supplier Thermo Scientific™ Supplier No. 042191
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Separate cationic and neutral species in a single run using this solvent-compatible mixed-mode column.

Separate cationic and neutral analytes in a single run by using Thermo Scientific™ OmniPac™ PCX-500 Columns. With cation-exchange and reversed-phase properties, this polymer-based mixed-mode column is solvent compatible and stable over the entire pH range of 1-14. Solvents may be used to affect selectivity as well as to remove contaminants to extend column lifetime.

This column is ideal for analyzing a wide range of compounds including adrenergics, alcohols, anilines, antidepressants, anti-inflammatory agents, plating bath brighteners, cephalosporins, stains and dyes, herbicides, nucleotides and nucleosides, peptides, sulfonamides, and water-soluble vitamins.
  • Macroporous hydrophobic core provides reversed-phase retention and is coated with an cation-exchange latex bead layer for cation-exchange retention
  • Ion exchange capacity of approximately 120 μeq/column
  • 100% solvent-compatible and should always be run with at least 1% organic solvent in the mobile phase
TRUSTED_SUSTAINABILITY

Specifications

Column Type Mixed Mode
Flow Rate 1 mL/min
For Use With (Application) Reversed Phase and Ion Exchange
Max. Pressure 4000 psi (275 bar)
Particle Size 8.5 μm
Stationary Phase Sulfonic Acid
Packing Material Spherical, Microporous Polymer
pH 0 to 14
Pore Size 1200 Å
Quantity Each
USP Type L50
Length (Metric) 250 mm
Diameter (Metric) 4 mm
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.