Learn More
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns
For the high-resolution separation and analysis of monoclonal antibody variants
Brand:  Thermo Scientific™ 088782
Additional Details : Weight : 0.65000kg
 View more versions of this product
Product Code. 15520244
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Unequalled high-resolution separations
  • Exceptionally high efficiency
  • Highest resolution of monoclonal antibody variants
  • Unmatched column-to-column and lot-to-lot reproducibility
  • Hydrophobic interactions eliminated
  • Excellent for stability testing
Specifications
Specifications
1 Lot of Resin
Bio-LC