Learn More
Thermo Scientific™ Dionex™ Fingertight Fitting with Double Ferrule
Product Code. 11332143 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Inert Valve, Older Generation Nano and Capillary LC Systems
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11332143 Inert Valve, Older Generation Nano and Capillary LC Systems Each
1 options
Product Code. 11332143 Supplier Thermo Scientific™ Supplier No. 160693
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Dionex Fingertight Fitting with Double Ferrules are for older generation nano and capillary LC systems.
Thermo Scientific™ Dionex Fingertight Fitting with Double Ferrules are for older generation nano and capillary LC systems.
TRUSTED_SUSTAINABILITY
Specifications
Description Fingertight fitting
For Use With (Equipment) Inert Valve, Older Generation Nano and Capillary LC Systems
Type Fingertight Fitting with Double Ferrule
Product Line Dionex