missing translation for 'onlineSavingsMsg'
Learn More

Diolein, TRC

Product Code. 30351615
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351615 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351615 Supplier Toronto Research Chemicals Supplier No. TRCD263785500MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glyceryl Dioleate
CAS 25637-84-7
Molecular Formula C39H72O5 • C39H72O5
Quantity 500 mg
Synonym DAG-18:1/18:1, DGO 8, Diolein, Emulsifier TS-PH 008, Glycerin Dioleate, Glycerol Dioleate, Glyceryl Dioleate, Ligalub 12GE, Nikkol DGO 80, Oleic Acid Diglyceride
InChI Formula InChI=1S/2C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h2*17-20,37,40H,3-16,21-36H2,1-2H3/b2*19-17-,20-18-/t;37-/m.0/s1
SMILES OC(COC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.OC[C@H](OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O
IUPAC Name (S)-3-hydroxypropane-1,2-diyl dioleate--2-hydroxypropane-1,3-diyl dioleate (1/1)
Molecular Weight (g/mol) 1242
Formula Weight 1241.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.