missing translation for 'onlineSavingsMsg'
Learn More

Diolein, > 99%, MP Biomedicals™

Product Code. 11481032 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481032 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11481032 Supplier MP Biomedicals Supplier No. 0219515191
  • ONLINE EXCLUSIVE PRICE
    £538.00
    £504.00 / 1g
    Save £34.00
    6% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25637-84-7
Molecular Formula C39H72O5
Synonym Glycerol-1, 2-and-1, 3-dioleate

Specifications

Color Colorless
Density 0.921g/mL
Assay Percent Range ∽ 99%
Quantity 1 g
Formula Weight 621
Percent Purity >99%
Physical Form Frozen Liquid
Chemical Name or Material Diolein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.