missing translation for 'onlineSavingsMsg'
Learn More

Dioctyltinmercaptide (~90%), TRC

Product Code. 30383164
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383164 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30383164 Supplier Toronto Research Chemicals Supplier No. TRCD4821502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Plastic additive 23
CAS 15571-58-1
Molecular Formula C36 H72 O4 S2 Sn
Quantity 2.5 mg
Synonym 10-Ethyl-4,4-dioctyl-7-oxo-8-oxa-3,5-dithia-4-stannatetradecanoic acid 2-ethylhexyl ester, [(Dioctylstannylene)dithio]diacetic acid bis(2-ethylhexyl) ester, Bis[(carboxymethyl)thio]dioctyltin bis(2-ethylhexyl) ester, 17MOK, Advastab 17MOK, Advastab 17MOL, Bis(2-ethylhexyl thioglycolato)dioctyltin, Bis(2-ethylhexyl) [(dioctylstannylene)dithio]diacetate, CAT 214, Dioctyltin bis(2-ethylhexyl thioglycolate), KS 2000A10, Reatinor 847, Thermolite 895, Plastic additive 23 (EP)
InChI Formula InChI=1S/2C10H20O2S.2C8H17.Sn/c2*1-3-5-6-9(4-2)7-12-10(11)8-13;2*1-3-5-7-8-6-4-2;/h2*9,13H,3-8H2,1-2H3;2*1,3-8H2,2H3;/q;;;;+2/p-2
SMILES CCCCCCCC[Sn](CCCCCCCC)(SCC(=O)OCC(CC)CCCC)SCC(=O)OCC(CC)CCCC
IUPAC Name 2-ethylhexyl 2-[[2-(2-ethylhexoxy)-2-oxoethyl]sulfanyl-dioctylstannyl]sulfanylacetate
Molecular Weight (g/mol) 751.79
Formula Weight 752.3894
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.