missing translation for 'onlineSavingsMsg'
Learn More

Dioctyldilauryltin ( >90%), TRC

Product Code. 30381627
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381627 100 mg
30370053 500 mg
30380465 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381627 Supplier Toronto Research Chemicals Supplier No. TRCD481773100MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3648-18-8
Molecular Formula C40H80O4Sn
Molecular Weight (g/mol) 743.77
Synonym dioctylbis[(1-oxododecyl)oxy]-Stannane, Dioctyltin dilaurate (6CI), Stannane, bis(lauroyloxy)dioctyl- (8CI), Tin, bis(lauroyloxy)dioctyl- (7CI), Dioctylbis[(1-oxododecyl)oxy]stannane, Lauric acid, dioctylstannylene deriv. (8CI), ADK Stab OT 1, DOTL, Di-n-octyltin dilaurate, Di-n-ocytyltin dilaurate, Dioctyldi(lauroyloxy)tin, Dioctyltin didodecanoate, Dioctyltindilaureate, Dioctytin dilaurate, KS 1200A1, LZ 082, Neostann U 810, OL 1, OT 1, OT 1 (stabilizer), Stann SNT, TIB KAT 216, Tegokat 216, Tin dioctyldilaurate, U 810
IUPAC Name [dodecanoyloxy(dioctyl)stannyl] dodecanoate
SMILES CCCCCCCCCCCC(=O)O[Sn](CCCCCCCC)(CCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C12H24O2.2C8H17.Sn/c2*1-2-3-4-5-6-7-8-9-10-11-12(13)14;2*1-3-5-7-8-6-4-2;/h2*2-11H2,1H3,(H,13,14);2*1,3-8H2,2H3;/q;;;;+2/p-2
Formula Weight 744.5079
Chemical Name or Material Dioctyldilauryltin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.