missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dioctyl Terepthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Dioctyl Terepthalate-d4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24H34D4O4 |
| Quantity | 5 mg |
| Synonym | ADK Cizer D 810-d4, Bis(2-ethylhexyl) Terephthalate-d4, DOTP-d4, Di-(2-ethylhexyl) terephthalate-d4, Dioctyl Terephthalate-d4, Eastman 168-d4, Eastman TM 168-d4, JP 110-d4, Kodaflex DOTP-d4, NEO-T-d4, Plasticizer 168-d4, 1,4-Benzenedicarboxylic Acid Bis(2-ethylhexyl) Ester-d4, Terephthalic Acid Bis(2-ethylhexyl) Ester-d4, DEHT-d4 |
| InChI Formula | InChI=1S/C24H38O4/c1-5-9-11-19(7-3)17-27-23(25)21-13-15-22(16-14-21)24(26)28-18-20(8-4)12-10-6-2/h13-16,19-20H,5-12,17-18H2,1-4H3/i13D,14D,15D,16D |
| SMILES | O=C(OCC(CC)CCCC)C1=C([2H])C([2H])=C(C(OCC(CC)CCCC)=O)C([2H])=C1[2H] |
| Molecular Weight (g/mol) | 394.58 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?