missing translation for 'onlineSavingsMsg'
Learn More

Dioctadecyl 3,3’-Thiodipropionate, TRC

Product Code. 30385317
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385317 Supplier Toronto Research Chemicals Supplier No. TRCD4811905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Plastic additive 17
CAS 693-36-7
Molecular Formula C42 H82 O4 S
Quantity 5 g
Synonym 3,3'-Thiobispropanoic acid 1,1'-dioctadecyl ester, 3,3'-Thiobispropanoic acid dioctadecyl ester, 3,3'-Thiodipropionic acid dioctadecyl ester, Advastab 802, Advastab PS 802, Antiox S, Antioxidant 802, Arbestab DSTDP, Carstab DSTDP, Cyanox OTDP, Cyanox STDP, DSTDP, DSTP, DSTP Yoshitomi, Dioctadecyl 3,3'-thiodipropionate, Dioctadecyl thiodipropionate, Distearyl 3,3'-thiodipropionate, Distearyl thiodipropionate, Distearyl β,β'-thiodipropionate, Distearyl β-thiodipropionate, Distearyl-3,3-thiodipropionate, Evanstab 18, Hostanox SE 2, Hostanox SE 4, Hostanox VP-SE 2, Irganox 802FD, Irganox PS 802, Irganox PS 802FD, Irganox PS 802FL, Lasumit SG, Lowinox DSTDP, Lusmit SS, NSC 65493, Naugard DSTDP, Negonox DSTP, PS 802, PS 802FL, Plastanox STDP, Plastanox STDP Antioxidant, Seenox DS, Songnox DSTDP, Stearyl 3,3'-thiodipropionate, Sumilizer TPS, Thio 1, Varox DSTDP, Yoshinox DSTDP, Yoshinox DSTP
InChI Formula InChI=1S/C42H82O4S/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37-45-41(43)35-39-47-40-36-42(44)46-38-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC
IUPAC Name octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate
Molecular Weight (g/mol) 683.16
Formula Weight 682.5934
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.