missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dioctadecyl 3,3’-Thiodipropionate, TRC
Specifications
Specifications
| Chemical Name or Material | Plastic additive 17 |
| CAS | 693-36-7 |
| Molecular Formula | C42 H82 O4 S |
| Quantity | 10 g |
| Synonym | 3,3'-Thiobispropanoic acid 1,1'-dioctadecyl ester, 3,3'-Thiobispropanoic acid dioctadecyl ester, 3,3'-Thiodipropionic acid dioctadecyl ester, Advastab 802, Advastab PS 802, Antiox S, Antioxidant 802, Arbestab DSTDP, Carstab DSTDP, Cyanox OTDP, Cyanox STDP, DSTDP, DSTP, DSTP Yoshitomi, Dioctadecyl 3,3'-thiodipropionate, Dioctadecyl thiodipropionate, Distearyl 3,3'-thiodipropionate, Distearyl thiodipropionate, Distearyl β,β'-thiodipropionate, Distearyl β-thiodipropionate, Distearyl-3,3-thiodipropionate, Evanstab 18, Hostanox SE 2, Hostanox SE 4, Hostanox VP-SE 2, Irganox 802FD, Irganox PS 802, Irganox PS 802FD, Irganox PS 802FL, Lasumit SG, Lowinox DSTDP, Lusmit SS, NSC 65493, Naugard DSTDP, Negonox DSTP, PS 802, PS 802FL, Plastanox STDP, Plastanox STDP Antioxidant, Seenox DS, Songnox DSTDP, Stearyl 3,3'-thiodipropionate, Sumilizer TPS, Thio 1, Varox DSTDP, Yoshinox DSTDP, Yoshinox DSTP |
| InChI Formula | InChI=1S/C42H82O4S/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37-45-41(43)35-39-47-40-36-42(44)46-38-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC |
| IUPAC Name | octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate |
| Molecular Weight (g/mol) | 683.16 |
| Formula Weight | 682.5934 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?