missing translation for 'onlineSavingsMsg'
Learn More

DIO3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18429357 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429357 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429357 Supplier Novus Biologicals Supplier No. NBP105767MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIO3 Polyclonal antibody specifically detects DIO3 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIO3
Applications Flow Cytometry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat DIO3 (within residues 250-300). [Swiss-Prot# P49897]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1735
Target Species Human, Mouse, Rat, Hamster, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.