missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05767JF646
This item is not returnable.
View return policy
Description
DIO3 Polyclonal specifically detects DIO3 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| DIO3 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| 5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5′ deiodinase, Type-III 5′-deiodinase | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 1735.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DIO3 | |
| Synthetic peptide made to an internal portion of rat DIO3 (within residues 250-300). [Swiss-Prot No. P49897] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Hamster, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction