missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05767H
This item is not returnable.
View return policy
Description
DIO3 Polyclonal specifically detects DIO3 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| DIO3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| 5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase | |
| Rabbit | |
| 31 kDa | |
| 0.1 mL | |
| Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 1735 | |
| Human, Mouse, Rat, Hamster, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HRP | |
| PBS with No Preservative | |
| DIO3 | |
| Synthetic peptide made to an internal portion of rat DIO3 (within residues 250-300). [Swiss-Prot# P49897] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, rat, hamster and mouse. Rabbit reactivity via customer review. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction