missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56543
This item is not returnable.
View return policy
Description
DIO2 Polyclonal specifically detects DIO2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| DIO2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| 5DII, D2, deiodinase, iodothyronine, type II, DIOII, EC 1.97.1, EC 1.97.1.10, ITDI2, SelY, TXDI2thyroxine deiodinase, type II, Type 2 DI, type 2 iodothyronine deiodinase, type II iodothyronine deiodinase, type-II 5'deiodinase, Type-II 5'-deiodinase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1734 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| DIO2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EWRRMLTSEGLRCVWKSFLLDAYKQVKLGEDAPNSSVVHVSSTEGGDNSGNGTQEKIAEGATCHLLDFASPERP | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion