missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dinotefuran, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Dinotefuran |
| CAS | 165252-70-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 H14 N4 O3 |
| Quantity | 100 mg |
| Synonym | Guanidine, N-methyl-N'-nitro-N''-[(tetrahydro-3-furanyl)methyl]- (9CI), N''-Methyl-N-nitro-N'-[(tetrahydro-3-furanyl)methyl]guanidine, Albarin, Alpine WSG, Dinotefuran, MTI 446, Mikeblock, Nidinotefuran, Safari, Safari (insecticide), Safari 20SG, Starkle, Tenchu, Venom (insecticide), Zylam |
| InChI Formula | InChI=1S/C7H14N4O3/c1-8-7(10-11(12)13)9-4-6-2-3-14-5-6/h6H,2-5H2,1H3,(H2,8,9,10) |
| SMILES | CN\C(=N/CC1CCOC1)\N[N+](=O)[O-] |
| IUPAC Name | 1-methyl-3-nitro-2-(oxolan-3-ylmethyl)guanidine |
| Molecular Weight (g/mol) | 202.21 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction