missing translation for 'onlineSavingsMsg'
Learn More

Dinoseb, TRC

Product Code. 30381740
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381740 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381740 Supplier Toronto Research Chemicals Supplier No. TRCD48049050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dinoseb
CAS 88-85-7
Purity Grade Notes HPLC
Molecular Formula C10 H12 N2 O5
Quantity 50 mg
Synonym Phenol, 2-(1-methylpropyl)-4,6-dinitro-, Phenol, 2-sec-butyl-4,6-dinitro- (6CI,8CI), 2-(1-Methylpropyl)-4,6-dinitrophenol, 2,4-Dinitro-6-(1-methylpropyl)phenol, 2,4-Dinitro-6-sec-butylphenol, 2-sec-Butyl-4,6-dinitrophenol, 4,6-Dinitro-2-(1-methyl-n-propyl)phenol, 4,6-Dinitro-2-(2-butyl)phenol, 4,6-Dinitro-2-sec-butylphenol, 4,6-Dinitro-o-sec-butylphenol, 6-sec-Butyl-2,4-dinitrophenol, Aatox, BNP 20, BNP 30, Blaartox, Butaphen, Butaphene, Chemox P.E., DBNF, DN 289, DNBP, DNOSBP, DNSBP, Desicoil, Dibutox, Dibutox 20CE, Dinoseb, Dow General, Dytop, Hivertox, Ladob, Laseb, Liro DNBP, NSC 202753, Premerge, Super Kabrol, WSX 8365
InChI Formula InChI=1S/C10H12N2O5/c1-3-6(2)8-4-7(11(14)15)5-9(10(8)13)12(16)17/h4-6,13H,3H2,1-2H3
SMILES CCC(C)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2-butan-2-yl-4,6-dinitrophenol
Molecular Weight (g/mol) 240.21
Formula Weight 240.0746
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.