missing translation for 'onlineSavingsMsg'
Learn More

Dinosam, TRC

Product Code. 30138522
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138522 500 mg 500mg
30003257 250 mg 250mg
30010975 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138522 Supplier Toronto Research Chemicals Supplier No. TRCD409730500MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-36-3
Molecular Formula C11 H14 N2 O5
Molecular Weight (g/mol) 254.2393
Synonym 2-(1-Methylbutyl)-4,6-dinitrophenol, 2,4-Dinitro-6-(1-methylbutyl)phenol, 2,4-Dinitro-6-(pentan-2-yl)phenol, 2-(1-Methyl-n-butyl)-4,6-dinitrophenol, 2-(2-Hydroxy-3,5-dinitrophenyl)pentane, 2-sec-Amyl-4,6-dinitrophenol, Chemox General, DNAP, DNOSAP, Dinosam, NSC 21493
IUPAC Name 2,4-dinitro-6-pentan-2-ylphenol
SMILES CCCC(C)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H14N2O5/c1-3-4-7(2)9-5-8(12(15)16)6-10(11(9)14)13(17)18/h5-7,14H,3-4H2,1-2H3
Formula Weight 254.0903
Percent Purity >95%
Chemical Name or Material Dinosam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.