missing translation for 'onlineSavingsMsg'
Learn More

Dinoprost tromethamine salt, MedChemExpress

Product Code. 16465448
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465448

Brand: Medchem Express HY12956A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dinoprost tromethamine salt (Prostaglandin F2α tromethamine salt) is an orally active, potent prostaglandin F (PGF) receptor (FP receptor) agonist. Dinoprost tromethamine salt is a luteolytic hormone produced locally in the endometrial luminal epithelium and corpus luteum (CL). Dinoprost tromethamine salt plays a key role in the onset and progression of labour.

Dinoprost tromethamine salt (Prostaglandin F2α tromethamine salt) is an orally active, potent prostaglandin F (PGF) receptor (FP receptor) agonist. Dinoprost tromethamine salt is a luteolytic hormone produced locally in the endometrial luminal epithelium and corpus luteum (CL). Dinoprost tromethamine salt plays a key role in the onset and progression of labour.

Specifications

Chemical Name or Material Dinoprost tromethamine salt
Physical Form Solid
CAS 38562-01-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H45NO8
Synonym Prostaglandin F2α tromethamine salt PGF2α THAM Prostaglandin F2α THAM
Solubility Information H2O : ≥ 100 mg/mL (210.25 mM) ∣DMSO : ≥ 100 mg/mL (210.25 mM)
SMILES CCCCC[C@H](O)/C=C/[C@H]1[C@H](O)C[C@H](O)[C@@H]1C/C=C\CCCC(O)=O.OCC(CO)(N)CO
Molecular Weight (g/mol) 475.62
Formula Weight 475.62
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.