missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIMT1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DIMT1L |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16874794
|
Proteintech
15563-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16864794
|
Proteintech
15563-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3′-end of 18S rRNA in the 40S particle.Specifications
| DIMT1L | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9D0D4, Q9UNQ2 | |
| 27292, 294718, 66254 | |
| DIMT1L Fusion Protein Ag7721 | |
| Primary | |
| -20°C | |
| DIMT1 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 18S rRNA dimethylase, DIM1, DIMT1, DIMT1L, HSA9761, HUSSY5 | |
| DIMT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title