missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117911
This item is not returnable.
View return policy
Description
DIMT1 Polyclonal Antibody for Western Blot, IHC (P), IP
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA in the 40S particle.
Specifications
| DIMT1 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9UNQ2 | |
| DIMT1 | |
| E. coli-derived Human DIMT1L fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DIMT1 | |
| 1500031M22Rik; 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; 18S rRNA dimethylase; AV111347; DIM1; DIM1 dimethyladenosine transferase 1 homolog; DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae); DIM1 dimethyladenosine transferase 1-like; DIM1 dimethyladenosine transferase 1-like (S. cerevisiae); dimethyladenosine transferase; Dimt1; Dimt1l; HSA9761; HUSSY-05; HUSSY5; probable 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; Probable 18S rRNA dimethylase; probable dimethyladenosine transferase; Probable S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase; RGD1311752; S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 27292 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction