missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560170
This item is not returnable.
View return policy
Description
Immunogen sequence: GGLMFNTGIG QHILKNPLII NSIIDKAALR PTDVVLEVGP GTGNMTVKLL EKAKKVVACE LDPRLVAELH KRVQGTPVAS KLQVLVGDVL KTDLPFFDTC V Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA in the 40S particle.
Specifications
| DIMT1 | |
| Polyclonal | |
| Unconjugated | |
| DIMT1 | |
| 1500031M22Rik; 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; 18S rRNA dimethylase; AV111347; DIM1; DIM1 dimethyladenosine transferase 1 homolog; DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae); DIM1 dimethyladenosine transferase 1-like; DIM1 dimethyladenosine transferase 1-like (S. cerevisiae); dimethyladenosine transferase; Dimt1; Dimt1l; HSA9761; HUSSY-05; HUSSY5; probable 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; Probable 18S rRNA dimethylase; probable dimethyladenosine transferase; Probable S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase; RGD1311752; S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UNQ2 | |
| DIMT1 | |
| Recombinant protein corresponding to Human DIMT1. Recombinant protein control fragment (Product #RP-91718). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction