missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ Diminazene aceturate

Product Code. 18725816
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18725816

Brand: Tocris™ 6705/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ASIC blocker; antihyperalgesic

Diminazene aceturate is an ASIC blocker (IC50 values are 202, ∽320, ∽320 and 864 nM for ASIC1b, ASIC1a, ASIC3 and ASIC2a, respectively). Exhibits no activity in ENaC channels expressed in oocytes. Accelerates desensitization of ASIC currents in hippocampal neurons. Inhibits mesotrypsin (Ki = 3.6?0.3 μM). Displays in vitro and in vivo anti-inflammatory effects in mice. Antihyperalgesic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-(1-Triazene-1,3-diyl)bis[benzenecarboximidamide] bis(N-acetylglycinate)
CAS 908-54-3
Quantity 100 mg
Molecular Formula C14H15N7.2C4H7NO3
Formula Weight 515.52
Product Type ASIC Blocker, Antihyperalgesic
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.