missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diminazene 1000 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Diminazene aceturate |
| CAS | 908-54-3 |
| Analyte or Component Names | Diminazene aceturate |
| Shipping Condition | +20°C |
| Molecular Formula | C14 H15 N7 . 2 C4 H7 N O3 |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Glycine, N-acetyl-, compd. with 4,4'-(1-triazene-1,3-diyl)bis[benzenecarboximidamide] (2:1), Benzamidine, 4,4'-diazoaminodi-, diaceturate (6CI), Glycine, N-acetyl-, compd. with 4,4'-(azoamino)dibenzamidine (7CI), Glycine, N-acetyl-, compd. with 4,4'-(diazoamino)dibenzamidine (2:1) (8CI), Benzamidine, 4,4'-(diazoamino)di-, compd. with N-acetylglycine (1:2) (8CI), Benzenecarboximidamide, 4,4'-(1-triazene-1,3-diyl)bis-, compd. with N-acetylglycine (1:2), 1,3-Bis(p-amidinophenyl)triazene bis(N-Acetylglycinate), 1,3-Bis[4-guanylphenyl]triazene diaceturate, 4,4'-(Diazoamino)dibenzamidine diaceturate, 4,4'-Diamidinodiazoaminobenzene diaceturate, Azidin, Azidine, Berenil, Beronal, Di-(4-amidinophenyl)-triazine-(N-1,3)-diaceturate, Diaminazene aceturate, Diminasan, Diminazene aceturate, Diminazene diaceturate, Diminazine aceturate, Diminazine diaceturate, Fatrybanil, Ganasag, Ganaseg, Tricip Plus, Veriben, p,p'-Diguanyldiazoaminobenzene diaceturate |
| InChI Formula | InChI=1S/C14H15N7.2C4H7NO3/c15-13(16)9-1-5-11(6-2-9)19-21-20-12-7-3-10(4-8-12)14(17)18;2*1-3(6)5-2-4(7)8/h1-8H,(H3,15,16)(H3,17,18)(H,19,20);2*2H2,1H3,(H,5,6)(H,7,8) |
| SMILES | CC(=O)NCC(=O)O.CC(=O)NCC(=O)O.NC(=N)c1ccc(NN=Nc2ccc(cc2)C(=N)N)cc1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction