missing translation for 'onlineSavingsMsg'
Learn More

Dimidium Bromide, MP Biomedicals

Product Code. 11436742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436742 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436742 Supplier MP Biomedicals Supplier No. 0215095880
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 518-67-2
Assay Percent Range ∽95%
Molecular Formula C20H18BrN3
MDL Number MFCD00011757
Quantity 100 mg
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
Molecular Weight (g/mol) 380.29
PubChem CID 68207
Formula Weight 380.3
Chemical Name or Material Dimidium Bromide, Fluorescent probe for nucleic acids; interchelating agent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.